Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCATCATGGTTTTTGAATGAAGC[C/A]TTCCACACATGCACGGGAACTATGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 137162 | ||||||||||||||||||||
Literature Links: |
GABRR2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GABRR2 - gamma-aminobutyric acid type A receptor rho2 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002043.4 | 2325 | Missense Mutation | AGG,ATG | R,M 369 | NP_002034.3 | |
XM_011535713.2 | 2325 | Missense Mutation | AGG,ATG | R,M 325 | XP_011534015.1 | |
XM_011535714.2 | 2325 | Missense Mutation | AGG,ATG | R,M 270 | XP_011534016.1 | |
XM_011535715.2 | 2325 | Missense Mutation | AGG,ATG | R,M 270 | XP_011534017.1 | |
XM_011535716.2 | 2325 | Missense Mutation | AGG,ATG | R,M 270 | XP_011534018.1 | |
XM_011535717.2 | 2325 | Missense Mutation | AGG,ATG | R,M 270 | XP_011534019.1 |