Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGAGAGAGAGGAAAATTCCCTAAC[C/T]CTTGGTTTAAAGACAATATTCATTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 107580 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TFAP2A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TFAP2A - transcription factor AP-2 alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001032280.2 | 2701 | UTR 3 | NP_001027451.1 | |||
NM_001042425.1 | 2701 | UTR 3 | NP_001035890.1 | |||
NM_003220.2 | 2701 | UTR 3 | NP_003211.1 | |||
XM_006715175.2 | 2701 | UTR 3 | XP_006715238.1 | |||
XM_011514833.2 | 2701 | UTR 3 | XP_011513135.1 | |||
XM_017011232.1 | 2701 | UTR 3 | XP_016866721.1 |