Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACAGTGGGTATTTGTCAGAACTAC[C/T]GTGCTGAGAAAAGGCACAGGAGACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
RNF182 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RNF182 - ring finger protein 182 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001165032.1 | Intron | NP_001158504.1 | ||||
NM_001165033.1 | Intron | NP_001158505.1 | ||||
NM_001165034.1 | Intron | NP_001158506.1 | ||||
NM_152737.3 | Intron | NP_689950.1 | ||||
XM_017010448.1 | Intron | XP_016865937.1 | ||||
XM_017010449.1 | Intron | XP_016865938.1 | ||||
XM_017010450.1 | Intron | XP_016865939.1 |