Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGGGGCTGTTGCCCTTGTGTCAT[C/T]CCCCCGCCTTTTAACTGCAAAGGTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
RBM24 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RBM24 - RNA binding motif protein 24 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143941.1 | 134 | UTR 5 | NP_001137413.1 | |||
NM_001143942.1 | 134 | Intron | NP_001137414.1 | |||
NM_153020.2 | 134 | Intron | NP_694565.1 | |||
XM_005248932.3 | 134 | Intron | XP_005248989.1 | |||
XM_011514387.2 | 134 | Intron | XP_011512689.1 | |||
XM_011514388.2 | 134 | Intron | XP_011512690.1 | |||
XM_011514389.2 | 134 | Intron | XP_011512691.1 |