Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTGTCATTCTGCCATTAGTGCAAT[G/T]TCAGTAACATTAAAGAGTTTCACGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611049 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC17A2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC17A2 - solute carrier family 17 member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286123.1 | Intron | NP_001273052.1 | ||||
NM_001286125.1 | Intron | NP_001273054.1 | ||||
NM_005835.3 | Intron | NP_005826.1 | ||||
XM_005248784.2 | Intron | XP_005248841.1 | ||||
XM_006714949.3 | Intron | XP_006715012.1 | ||||
XM_006714950.2 | Intron | XP_006715013.1 | ||||
XM_006714951.1 | Intron | XP_006715014.1 | ||||
XM_017010159.1 | Intron | XP_016865648.1 | ||||
XM_017010160.1 | Intron | XP_016865649.1 |