Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGTGACCAAGGCACAGAAGAAGGA[C/T]GGCAAGAAGCGCAAGCGCAGCCGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602807 MIM: 602815 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HIST1H2BI PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HIST1H2BI - histone cluster 1, H2bi | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003525.2 | 78 | Silent Mutation | GAC,GAT | D,D 26 | NP_003516.1 |
HIST1H3G - histone cluster 1, H3g | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |