Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTTGTAGCTTTGAGATTCAGACT[C/T]GTTTGCACACAAGGCTTTTATGATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608196 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MYLK4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MYLK4 - myosin light chain kinase family member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005249078.3 | Intron | XP_005249135.2 | ||||
XM_005249079.2 | Intron | XP_005249136.1 | ||||
XM_006715082.3 | Intron | XP_006715145.1 | ||||
XM_011514567.2 | Intron | XP_011512869.1 | ||||
XM_017010819.1 | Intron | XP_016866308.1 |
WRNIP1 - Werner helicase interacting protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020135.2 | Intron | NP_064520.2 | ||||
NM_130395.2 | Intron | NP_569079.1 | ||||
XM_005249232.3 | Intron | XP_005249289.1 | ||||
XM_017011060.1 | Intron | XP_016866549.1 |