Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600214 MIM: 603099 MIM: 164951 MIM: 176311 MIM: 602677 | ||||||||||||||||||||
Literature Links: |
AGER PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AGER - advanced glycosylation end product-specific receptor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
AGPAT1 - 1-acylglycerol-3-phosphate O-acyltransferase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GPSM3 - G-protein signaling modulator 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR6833 - microRNA 6833 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NOTCH4 - notch 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PBX2 - PBX homeobox 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002586.4 | Intron | NP_002577.2 |
RNF5 - ring finger protein 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |