Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAGAGGTCCTAGTGAAGATTGTAC[A/G]TTAACTCTAGGTGGGAAGGATGGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 147267 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ITPR3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ITPR3 - inositol 1,4,5-trisphosphate receptor type 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002224.3 | Intron | NP_002215.2 | ||||
XM_011514577.2 | Intron | XP_011512879.1 | ||||
XM_017010832.1 | Intron | XP_016866321.1 |
LOC101929188 - uncharacterized LOC101929188 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |