Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAGGGTCCAGCTCAGTTAAAGGAA[G/T]AATTTTCTCCTGGCCAGAGTTGGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604100 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GRM4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GRM4 - glutamate metabotropic receptor 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000841.3 | Intron | NP_000832.1 | ||||
NM_001256809.2 | Intron | NP_001243738.1 | ||||
NM_001256811.2 | Intron | NP_001243740.1 | ||||
NM_001256812.2 | Intron | NP_001243741.1 | ||||
NM_001256813.2 | Intron | NP_001243742.1 | ||||
NM_001282847.1 | Intron | NP_001269776.1 | ||||
XM_017010790.1 | Intron | XP_016866279.1 | ||||
XM_017010791.1 | Intron | XP_016866280.1 | ||||
XM_017010792.1 | Intron | XP_016866281.1 | ||||
XM_017010793.1 | Intron | XP_016866282.1 |