Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTATTCTCATGCCCATTTCCCACCA[C/T]GGCCACCCTCAGGCCACACCAGCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612217 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C6orf106 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C6orf106 - chromosome 6 open reading frame 106 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022758.5 | 3899 | UTR 3 | NP_073595.2 | |||
NM_024294.3 | 3899 | UTR 3 | NP_077270.1 | |||
XM_005249298.2 | 3899 | Intron | XP_005249355.1 |