Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACTTAAAACCAGATATAAGAAACA[A/G]CCTAAGTCTTAGCAACTTCAGGCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600772 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105375030 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC105375030 - uncharacterized LOC105375030 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TAF11 - TATA-box binding protein associated factor 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270488.1 | 1780 | UTR 3 | NP_001257417.1 | |||
NM_005643.3 | 1780 | UTR 3 | NP_005634.1 | |||
XM_011514827.2 | 1780 | UTR 3 | XP_011513129.1 |
UHRF1BP1 - UHRF1 binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017754.3 | 1780 | Intron | NP_060224.3 |