Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGTCCTAATTACTCCTTGGGCTT[C/T]ACCGTGATGTGATCGCAGACATCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605255 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ETV7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ETV7 - ETS variant 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PXT1 - peroxisomal, testis specific 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152990.3 | 1109 | UTR 3 | NP_694535.2 | |||
XM_011514400.2 | 1109 | UTR 3 | XP_011512702.1 |