Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCTATCAAAAAGAAAAGAAGATAC[C/T]TAAAGTGCAGATATACAAGCACCGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611685 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RNF8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RNF8 - ring finger protein 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003958.3 | Intron | NP_003949.1 | ||||
NM_183078.2 | Intron | NP_898901.1 | ||||
XM_006715241.2 | Intron | XP_006715304.1 | ||||
XM_006715242.2 | Intron | XP_006715305.1 | ||||
XM_017011462.1 | Intron | XP_016866951.1 | ||||
XM_017011463.1 | Intron | XP_016866952.1 | ||||
XM_017011464.1 | Intron | XP_016866953.1 |