Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGTCGGGTGATATTCCGTTTGTTA[A/G]GATGGCTCTGTGATCATCAGAGATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607370 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KCNK17 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KCNK17 - potassium two pore domain channel subfamily K member 17 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135111.1 | 1650 | UTR 3 | NP_001128583.1 | |||
NM_031460.3 | 1650 | UTR 3 | NP_113648.2 |