Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGTCTTGTCACAATCCCCTTCGAG[C/T]CATGCCACAGGGCGTGGGCAACTAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608024 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C6orf201 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C6orf201 - chromosome 6 open reading frame 201 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001085401.2 | Intron | NP_001078870.1 | ||||
XM_005249112.3 | Intron | XP_005249169.1 | ||||
XM_011514613.2 | Intron | XP_011512915.1 | ||||
XM_017010861.1 | Intron | XP_016866350.1 |
ECI2 - enoyl-CoA delta isomerase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166010.1 | Intron | NP_001159482.1 | ||||
NM_006117.2 | Intron | NP_006108.2 | ||||
NM_206836.2 | Intron | NP_996667.2 | ||||
XM_006714957.1 | Intron | XP_006715020.1 | ||||
XM_017010170.1 | Intron | XP_016865659.1 |