Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACATTTTACCTGGTGGCAATGGGGA[A/C]CAGAAAGAAAAATAAAACAAAGAGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
DEFB133 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DEFB133 - defensin beta 133 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166478.1 | 43 | Missense Mutation | GTC,TTC | V,F 15 | NP_001159950.1 | |
XM_017010860.1 | 43 | Missense Mutation | GTC,TTC | V,F 15 | XP_016866349.1 |