Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGAGGAGAAGCAAGTCTTGGAAGA[C/T]GTAGAGTTTAGTTCTTAACATGAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 176636 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRIM2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PRIM2 - primase (DNA) subunit 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000947.4 | Intron | NP_000938.2 | ||||
NM_001282487.1 | Intron | NP_001269416.1 | ||||
NM_001282488.1 | Intron | NP_001269417.1 | ||||
XM_011514731.2 | Intron | XP_011513033.1 | ||||
XM_017011004.1 | Intron | XP_016866493.1 | ||||
XM_017011005.1 | Intron | XP_016866494.1 | ||||
XM_017011006.1 | Intron | XP_016866495.1 | ||||
XM_017011007.1 | Intron | XP_016866496.1 |