Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTATCTGGGTCTATCTGTCTATATA[C/T]GATTTGTCCATCTTTTTTTTTCCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613738 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AGMO PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AGMO - alkylglycerol monooxygenase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001004320.1 | Intron | NP_001004320.1 | ||||
XM_006715730.1 | Intron | XP_006715793.1 | ||||
XM_006715731.3 | Intron | XP_006715794.1 | ||||
XM_011515402.2 | Intron | XP_011513704.1 | ||||
XM_017012204.1 | Intron | XP_016867693.1 |