Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGGGAGACCAGGGTGCCCATCTT[C/T]CCAAACTGGGTCACCACCACCAGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
PSMG3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PSMG3 - proteasome assembly chaperone 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134340.1 | 774 | Intron | NP_001127812.1 | |||
NM_032302.3 | 774 | Silent Mutation | GGA,GGG | G,G 40 | NP_115678.1 |
PSMG3-AS1 - PSMG3 antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM184A - transmembrane protein 184A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |