Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTCCAGCTCTAGGAATCCAACCAA[G/T]AACAGGTCTTGTTTCTTCATTGTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616071 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C7orf31 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C7orf31 - chromosome 7 open reading frame 31 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_138811.3 | 1028 | Missense Mutation | ATT,CTT | I,L 526 | NP_620166.3 | |
XM_005249619.3 | 1028 | Missense Mutation | ATT,CTT | I,L 526 | XP_005249676.1 | |
XM_005249620.3 | 1028 | Missense Mutation | ATT,CTT | I,L 526 | XP_005249677.1 | |
XM_011515125.2 | 1028 | Missense Mutation | ATT,CTT | I,L 316 | XP_011513427.1 |