Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTTAATGCCTGAGAAATGCACTT[C/T]AGGGGTAGACTTATACCTTAAGTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRR15 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PRR15 - proline rich 15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_175887.2 | 2326 | UTR 3 | NP_787083.1 | |||
XM_005249653.3 | 2326 | UTR 3 | XP_005249710.1 | |||
XM_011515198.1 | 2326 | UTR 3 | XP_011513500.1 | |||
XM_011515199.1 | 2326 | UTR 3 | XP_011513501.1 | |||
XM_017011842.1 | 2326 | UTR 3 | XP_016867331.1 |