Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGAAAACTGCACAGTACAAACAC[A/T]GATACTGTTGACTCTTTTGCCAATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600418 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AMPH PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AMPH - amphiphysin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001635.3 | 3348 | UTR 3 | NP_001626.1 | |||
NM_139316.2 | 3348 | UTR 3 | NP_647477.1 | |||
XM_006715689.3 | 3348 | UTR 3 | XP_006715752.1 | |||
XM_006715690.3 | 3348 | UTR 3 | XP_006715753.1 | |||
XM_006715691.3 | 3348 | Intron | XP_006715754.1 | |||
XM_011515271.2 | 3348 | UTR 3 | XP_011513573.1 | |||
XM_017011995.1 | 3348 | UTR 3 | XP_016867484.1 | |||
XM_017011996.1 | 3348 | UTR 3 | XP_016867485.1 |
TRG-AS1 - T cell receptor gamma locus antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |