Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTTTAAGTCCTATTTTGCTATTT[A/G]ACCCTTGTTTATACTTTTGTTCAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105375240 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC105375240 - uncharacterized LOC105375240 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
YAE1D1 - Yae1 domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282446.1 | Intron | NP_001269375.1 | ||||
NM_020192.4 | Intron | NP_064577.1 |