Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATCCTCCTCTGAAAGGTTAAAATT[A/G]TATTAAAATTGTGCTGGGCATTATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 179550 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RALA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RALA - RALA Ras like proto-oncogene A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005402.3 | Intron | NP_005393.2 | ||||
XM_006715762.2 | Intron | XP_006715825.1 | ||||
XM_011515466.1 | Intron | XP_011513768.1 | ||||
XM_011515467.1 | Intron | XP_011513769.1 |