Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGTCCTTGAGGTAGGCGCCTTTCT[G/T]CCATTCAGACTTGAGTTCCAGCCAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611335 MIM: 611325 | ||||||||||||||||||||
Literature Links: |
SNORA5A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SNORA5A - small nucleolar RNA, H/ACA box 5A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORA5B - small nucleolar RNA, H/ACA box 5B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORA5C - small nucleolar RNA, H/ACA box 5C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TBRG4 - transforming growth factor beta regulator 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001261834.1 | 1659 | Missense Mutation | AAG,CAG | K,Q 623 | NP_001248763.1 | |
NM_004749.3 | 1659 | Missense Mutation | AAG,CAG | K,Q 612 | NP_004740.2 | |
NM_030900.3 | 1659 | Missense Mutation | AAG,CAG | K,Q 502 | NP_112162.1 | |
NM_199122.2 | 1659 | Missense Mutation | AAG,CAG | K,Q 502 | NP_954573.1 |