Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCTGACAACAGGCGCTGACCAAG[G/T]GGGCAGAGCAGCTGCACTCAGAGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611335 MIM: 611325 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SNORA5A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SNORA5A - small nucleolar RNA, H/ACA box 5A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORA5B - small nucleolar RNA, H/ACA box 5B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORA5C - small nucleolar RNA, H/ACA box 5C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TBRG4 - transforming growth factor beta regulator 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001261834.1 | Intron | NP_001248763.1 | ||||
NM_004749.3 | Intron | NP_004740.2 | ||||
NM_030900.3 | Intron | NP_112162.1 | ||||
NM_199122.2 | Intron | NP_954573.1 |