Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTGGCGGGCGCGCTGGCGCTGGC[C/G]GAGCTGCTGGGCGCGCCGGGGCCCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 103072 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADCY1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADCY1 - adenylate cyclase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001281768.1 | 252 | Intron | NP_001268697.1 | |||
NM_021116.2 | 252 | Silent Mutation | GCC,GCG | A,A 78 | NP_066939.1 | |
XM_005249584.3 | 252 | Silent Mutation | GCC,GCG | A,A 78 | XP_005249641.1 | |
XM_005249585.2 | 252 | Silent Mutation | GCC,GCG | A,A 78 | XP_005249642.1 |