Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAAATTCCATTCTGTTCCACATCA[A/G]CTTATGCATTGGAAATTTTGTATCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 164757 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BRAF PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BRAF - B-Raf proto-oncogene, serine/threonine kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004333.4 | 2402 | Intron | NP_004324.2 | |||
XM_005250045.2 | 2402 | UTR 3 | XP_005250102.1 | |||
XM_017012558.1 | 2402 | UTR 3 | XP_016868047.1 | |||
XM_017012559.1 | 2402 | Silent Mutation | AGC,AGT | S,S 798 | XP_016868048.1 |