Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCATCTTTTAGTGAAGAAAATAA[C/T]TGAACATGGTTTTCTTCTGAAGACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615133 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GALNTL5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GALNTL5 - polypeptide N-acetylgalactosaminyltransferase-like 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145292.3 | 237 | Intron | NP_660335.2 | |||
XM_006715862.3 | 237 | UTR 5 | XP_006715925.1 | |||
XM_011515849.2 | 237 | Intron | XP_011514151.1 | |||
XM_011515852.2 | 237 | Intron | XP_011514154.1 | |||
XM_011515853.2 | 237 | Intron | XP_011514155.1 | |||
XM_017011793.1 | 237 | Intron | XP_016867282.1 | |||
XM_017011794.1 | 237 | Intron | XP_016867283.1 | |||
XM_017011795.1 | 237 | Intron | XP_016867284.1 | |||
XM_017011796.1 | 237 | Intron | XP_016867285.1 |