Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCGGTTAGAGGGAGTTCTCATTG[A/G]AGATTTGTCTCTGGGATTAATGAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616252 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TCAF2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TCAF2 - TRPM8 channel associated factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130025.1 | 3626 | Intron | NP_001123497.1 | |||
NM_001130026.2 | 3626 | UTR 3 | NP_001123498.2 | |||
NM_173678.2 | 3626 | Intron | NP_775949.2 | |||
XM_006715927.3 | 3626 | UTR 3 | XP_006715990.1 | |||
XM_006715928.3 | 3626 | Intron | XP_006715991.1 | |||
XM_011516059.2 | 3626 | Intron | XP_011514361.1 |