Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGGATGTTCACAGGTGGGGTGCT[A/G]TTGGAGTAGTGTCCACTCTTGGCGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601679 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GTF2I PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GTF2I - general transcription factor IIi | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001163636.2 | Intron | NP_001157108.1 | ||||
NM_001280800.1 | Intron | NP_001267729.1 | ||||
NM_001518.4 | Intron | NP_001509.3 | ||||
NM_032999.3 | Intron | NP_127492.1 | ||||
NM_033000.3 | Intron | NP_127493.1 | ||||
NM_033001.3 | Intron | NP_127494.1 |