Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCACTACTGCTACCACCACTATCA[A/T]TACAGTCATTCCTATAATAAGAATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604918 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PCLO PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PCLO - piccolo presynaptic cytomatrix protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014510.2 | 19169 | Intron | NP_055325.2 | |||
NM_033026.5 | 19169 | UTR 3 | NP_149015.2 | |||
XM_017012006.1 | 19169 | UTR 3 | XP_016867495.1 | |||
XM_017012007.1 | 19169 | Intron | XP_016867496.1 |