Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGCCCAAGGGGGAGCGAGCCGGT[G/T]CTGCTGCAGGCTGAGGCTGCGGCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606090 MIM: 616403 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CROT PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CROT - carnitine O-octanoyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143935.1 | 13 | UTR 5 | NP_001137407.1 | |||
NM_001243745.1 | 13 | UTR 5 | NP_001230674.1 | |||
NM_021151.3 | 13 | UTR 5 | NP_066974.2 | |||
XM_011516337.2 | 13 | UTR 5 | XP_011514639.1 | |||
XM_017012370.1 | 13 | UTR 5 | XP_016867859.1 |
TP53TG1 - TP53 target 1 (non-protein coding) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |