Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAAACAAACAAACAAAAACTTATT[C/T]GTCATCGTAGGCTTGGAGCACATAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM133B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM133B - family with sequence similarity 133 member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040057.2 | Intron | NP_001035146.1 | ||||
NM_001288584.1 | Intron | NP_001275513.1 | ||||
NM_152789.3 | Intron | NP_690002.2 | ||||
XM_011516004.2 | Intron | XP_011514306.1 | ||||
XM_011516005.2 | Intron | XP_011514307.1 |