Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGCCCAGGGAAAGACAGGATACC[C/T]TGGGTCGGCTCCTCCACGTGACCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614133 MIM: 616465 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HEPACAM2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HEPACAM2 - HEPACAM family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039372.2 | 6 | Intron | NP_001034461.1 | |||
NM_001288804.1 | 6 | Intron | NP_001275733.1 | |||
NM_001288810.1 | 6 | Intron | NP_001275739.1 | |||
NM_198151.2 | 6 | Intron | NP_937794.1 | |||
XM_011516000.2 | 6 | Intron | XP_011514302.1 | |||
XM_011516001.1 | 6 | Intron | XP_011514303.1 |
VPS50 - VPS50, EARP/GARPII complex subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001257998.1 | 6 | UTR 5 | NP_001244927.1 | |||
NM_017667.3 | 6 | UTR 5 | NP_060137.2 | |||
NM_024553.2 | 6 | UTR 5 | NP_078829.1 | |||
XM_011516395.2 | 6 | Intron | XP_011514697.1 | |||
XM_011516396.2 | 6 | UTR 5 | XP_011514698.1 | |||
XM_017012412.1 | 6 | Intron | XP_016867901.1 |