Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCAGTCTATCACTCCGCTGTAAGT[A/G]GTTTGGCCAGCCCGGGCAGCCACCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603859 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC25A13 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC25A13 - solute carrier family 25 member 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001160210.1 | 1748 | Silent Mutation | ACC,ACT | T,T 558 | NP_001153682.1 | |
NM_014251.2 | 1748 | Silent Mutation | ACC,ACT | T,T 557 | NP_055066.1 | |
XM_006715831.3 | 1748 | Silent Mutation | ACC,ACT | T,T 568 | XP_006715894.1 | |
XM_011515727.2 | 1748 | Intron | XP_011514029.1 | |||
XM_017011663.1 | 1748 | Silent Mutation | ACC,ACT | T,T 554 | XP_016867152.1 | |
XM_017011664.1 | 1748 | Silent Mutation | ACC,ACT | T,T 273 | XP_016867153.1 | |
XM_017011665.1 | 1748 | Silent Mutation | ACC,ACT | T,T 273 | XP_016867154.1 |