Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGATTGTTTTTCAGTGGACTTCTG[A/G]GTTGGTTTCATCATTTCTTGTATAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614107 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KPNA7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KPNA7 - karyopherin subunit alpha 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_011516215.2 | Intron | XP_011514517.1 | ||||
XM_017012208.1 | Intron | XP_016867697.1 | ||||
XM_017012209.1 | Intron | XP_016867698.1 | ||||
XM_017012210.1 | Intron | XP_016867699.1 | ||||
XM_017012211.1 | Intron | XP_016867700.1 | ||||
XM_017012212.1 | Intron | XP_016867701.1 | ||||
XM_017012213.1 | Intron | XP_016867702.1 | ||||
XM_017012214.1 | Intron | XP_016867703.1 | ||||
XM_017012215.1 | Intron | XP_016867704.1 | ||||
XM_017012216.1 | Intron | XP_016867705.1 |