Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGCTTGGTCAGGGCAGCACTCTGG[A/C]TGAAGCTTTTCCCACATTCTTGGCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611272 | ||||||||||||||||||||
Literature Links: |
ZKSCAN5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZKSCAN5 - zinc finger with KRAB and SCAN domains 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF394 - zinc finger protein 394 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032164.2 | 1269 | Missense Mutation | AGC,ATC | S,I 396 | NP_115540.2 | |
XM_017012710.1 | 1269 | Intron | XP_016868199.1 | |||
XM_017012711.1 | 1269 | UTR 3 | XP_016868200.1 |
ZNF789 - zinc finger protein 789 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |