Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCACCTCCAGGGCCACATCCTGGAG[C/T]CAAGGGCTGTAGCCTGGGGATTCTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 158371 | ||||||||||||||||||||
Literature Links: |
MUC3A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MUC3A - mucin 3A, cell surface associated | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005960.1 | Intron | NP_005951.1 | ||||
XM_011516246.2 | Intron | XP_011514548.1 | ||||
XM_011516247.2 | Intron | XP_011514549.1 | ||||
XM_011516249.2 | Intron | XP_011514551.1 | ||||
XM_011516250.2 | Intron | XP_011514552.1 | ||||
XM_017012231.1 | Intron | XP_016867720.1 |