Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACTACATGGATGCCATGGCTGAGA[A/G]GGTCAGTGAGCCAGAAGGCTGGCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC100630923 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC100630923 - LOC100289561-PRKRIP1 readthrough | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR548O - microRNA 548o | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRKRIP1 - PRKR interacting protein 1 (IL11 inducible) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024653.3 | 360 | Missense Mutation | AAG,AGG | K,R 102 | NP_078929.1 |