Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613302 MIM: 615167 MIM: 604150 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ALKBH4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ALKBH4 - alkB homolog 4, lysine demethylase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LRWD1 - leucine rich repeats and WD repeat domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317721.1 | 853 | Intron | NP_001304650.1 | |||
NM_152892.2 | 853 | Intron | NP_690852.1 | |||
XM_017011845.1 | 853 | Intron | XP_016867334.1 |
MIR4467 - microRNA 4467 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR5090 - microRNA 5090 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
POLR2J - RNA polymerase II subunit J | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006234.4 | 853 | UTR 3 | NP_006225.1 | |||
XM_005250452.4 | 853 | Intron | XP_005250509.1 | |||
XM_017012353.1 | 853 | Intron | XP_016867842.1 |
RASA4B - RAS p21 protein activator 4B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |