Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAATTCATTAAGAGTTCTGATAAAT[C/G]TACTTCCTTACAGAATAGGTAAAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605502 MIM: 603131 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNAJC2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DNAJC2 - DnaJ heat shock protein family (Hsp40) member C2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001129887.1 | 2711 | Intron | NP_001123359.1 | |||
NM_014377.1 | 2711 | Intron | NP_055192.1 | |||
XM_005250269.2 | 2711 | Intron | XP_005250326.1 | |||
XM_011516028.2 | 2711 | Intron | XP_011514330.1 | |||
XM_011516029.2 | 2711 | Intron | XP_011514331.1 | |||
XM_011516030.2 | 2711 | Intron | XP_011514332.1 | |||
XM_017011968.1 | 2711 | Intron | XP_016867457.1 |
PMPCB - peptidase, mitochondrial processing beta subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004279.2 | 2711 | UTR 3 | NP_004270.2 | |||
XM_005250717.3 | 2711 | Intron | XP_005250774.1 | |||
XM_006716181.3 | 2711 | Intron | XP_006716244.1 |