Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATGAACTCAGGCGTTAGAATGTGG[G/T]GTTTAAACAGAGTCAAGAATTTAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602331 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ORC5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ORC5 - origin recognition complex subunit 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002553.3 | Intron | NP_002544.1 | ||||
NM_181747.3 | Intron | NP_859531.1 | ||||
XM_011516273.2 | Intron | XP_011514575.1 |