Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCACTGGAGAATGACATGTGGAAC[A/G]TGGCTTATTTACCTCCAATTCTTAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608444 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KMT2E PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KMT2E - lysine methyltransferase 2E | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018682.3 | Intron | NP_061152.3 | ||||
NM_182931.2 | Intron | NP_891847.1 | ||||
XM_005250493.1 | Intron | XP_005250550.1 | ||||
XM_011516400.1 | Intron | XP_011514702.1 | ||||
XM_017012435.1 | Intron | XP_016867924.1 |