Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTGTGATTTGTTTTAACTGCTCA[G/T]GAATAAGCAATACTTTAGCTTGACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601116 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GRM8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GRM8 - glutamate metabotropic receptor 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000845.2 | 3825 | UTR 3 | NP_000836.2 | |||
NM_001127323.1 | 3825 | UTR 3 | NP_001120795.1 | |||
XM_006715938.3 | 3825 | UTR 3 | XP_006716001.1 | |||
XM_011516091.1 | 3825 | UTR 3 | XP_011514393.1 | |||
XM_011516092.2 | 3825 | UTR 3 | XP_011514394.1 | |||
XM_011516094.1 | 3825 | UTR 3 | XP_011514396.1 | |||
XM_011516095.2 | 3825 | UTR 3 | XP_011514397.1 | |||
XM_011516101.2 | 3825 | UTR 3 | XP_011514403.1 | |||
XM_011516102.2 | 3825 | UTR 3 | XP_011514404.1 | |||
XM_011516103.2 | 3825 | UTR 3 | XP_011514405.1 | |||
XM_011516108.2 | 3825 | UTR 3 | XP_011514410.1 | |||
XM_017012074.1 | 3825 | UTR 3 | XP_016867563.1 | |||
XM_017012075.1 | 3825 | UTR 3 | XP_016867564.1 | |||
XM_017012076.1 | 3825 | UTR 3 | XP_016867565.1 | |||
XM_017012077.1 | 3825 | UTR 3 | XP_016867566.1 | |||
XM_017012078.1 | 3825 | UTR 3 | XP_016867567.1 | |||
XM_017012079.1 | 3825 | UTR 3 | XP_016867568.1 | |||
XM_017012080.1 | 3825 | UTR 3 | XP_016867569.1 |