Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAGGAATCTATATGGACCACATAA[A/G]ACTACAAATATTCATAAGACAGATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601082 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
UBE2H PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
UBE2H - ubiquitin conjugating enzyme E2 H | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001202498.1 | Intron | NP_001189427.1 | ||||
NM_003344.3 | Intron | NP_003335.1 | ||||
NM_182697.2 | Intron | NP_874356.1 |