Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTTTCTGCTCTGAAGGACATTATA[A/G]TATCATGAGAACAAAGATGAATATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605623 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LINC-PINT PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LINC-PINT - long intergenic non-protein coding RNA, p53 induced transcript | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MKLN1 - muskelin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145354.1 | Intron | NP_001138826.1 | ||||
NM_001321316.1 | Intron | NP_001308245.1 | ||||
NM_013255.4 | Intron | NP_037387.2 | ||||
XM_006715993.2 | Intron | XP_006716056.1 | ||||
XM_011516224.2 | Intron | XP_011514526.1 |