Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAAGTAGGAAATAATAAAGGGAAG[A/G]CATGATTGATACGCTGTCCTATAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607270 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AUTS2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AUTS2 - autism susceptibility candidate 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127231.2 | Intron | NP_001120703.1 | ||||
NM_001127232.2 | Intron | NP_001120704.1 | ||||
NM_015570.3 | Intron | NP_056385.1 | ||||
XM_005250257.2 | Intron | XP_005250314.1 | ||||
XM_011516010.2 | Intron | XP_011514312.1 | ||||
XM_011516011.2 | Intron | XP_011514313.1 | ||||
XM_011516012.2 | Intron | XP_011514314.1 | ||||
XM_011516013.2 | Intron | XP_011514315.1 | ||||
XM_011516014.2 | Intron | XP_011514316.1 | ||||
XM_011516017.2 | Intron | XP_011514319.1 | ||||
XM_011516018.2 | Intron | XP_011514320.1 | ||||
XM_017011951.1 | Intron | XP_016867440.1 |