Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATAATACACAGTTTCATGCTTCAG[G/T]TGAGCCAAGAACAACCTTCCCATTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605239 MIM: 611700 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATP6V0A4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ATP6V0A4 - ATPase H+ transporting V0 subunit a4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020632.2 | 2872 | UTR 3 | NP_065683.2 | |||
NM_130840.2 | 2872 | UTR 3 | NP_570855.2 | |||
NM_130841.2 | 2872 | UTR 3 | NP_570856.2 | |||
XM_005250393.1 | 2872 | UTR 3 | XP_005250450.1 | |||
XM_005250394.3 | 2872 | UTR 3 | XP_005250451.1 |
SVOPL - SVOP like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |